missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Growth Serum (U.S.)
Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
£204.00 - £262.00
Specifications
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
---|---|
Product Type | Bovine Growth Serum |
Quantity | 500 mL |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
12300003
|
Cytiva
SH30541.03HI |
Bovine Growth Serum | 500 mL | Heat Inactivated |
£204.00
500mL |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10453302
|
Cytiva
SH30541.03 |
Bovine Growth Serum (U.S.) | 500 mL | None |
£262.00
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Filtered through sequential 100nm (0.1μm) pore-size rated filters
Specifications
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
500 mL | |
Sequential-filtered, 100 nm | |
≤ 10 mg/dL |
Bovine Growth Serum | |
Sterile | |
≤ 10 EU/mL |