missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Serum Albumin (BSA)

Support cell culture growth with HyClone™ Bovine Serum Albumin (BSA).

£76.26 - £366.00

Specifications

Certifications/Compliance Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage 2° to 8° C
Form Powder
Sterility Non-sterile
Endotoxin Level ≤ 10 EU/mg
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10773877
missing translation for 'viewDocuments' Promotion Details
Cytiva
SH30574.01
Bovine Serum Albumin (BSA), Cell Culture Grade, pH 7.0, Lyophilized Powder. 10g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12330023
missing translation for 'viewDocuments' Promotion Details
Cytiva
SH30574.02
Bovine Serum Albumin (BSA), Cell Culture Grade, pH 7.0, Lyophilized Powder, 100g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

  • pH 7.0
  • Lyophilized powder
Specifications

Specifications

Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Powder
≤ 10 EU/mg
2° to 8° C
Non-sterile
6.5 to 7.5
Videos
SDS
Documents

Documents

Certificates