Learn More
Cytiva HyClone™ CDM4HEK293 Powdered Media
Promotes adenovirus and recombinant protein production in HEK 293 suspension cell culture.
Brand: Cytiva SH30859.01
Code : 83
Additional Details : CAS Number : 72-18-4 Weight : 0.11400kg
Description
- Optimized for high cell growth and high recombinant protein and vector production with human embryonic kidney (HEK) 293 cells
- Chemically defined, protein-free and animal-derived component-free medium available in liquid and powder form
- Designed for small- and large-scale biomanufacturing processes
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
- Supports direct or sequential adaptation
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No HEPES, No Phenol Red, No Sodium Bicarbonate |