missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4MAb™ Media
£102.00
Specifications
Content And Storage | 2° to 8°C, away from light |
---|---|
Form | Liquid |
Product Type | CDM4MAb Liquid Medium |
Sterilization Method | 0.2 μm Sterile-filtered |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10771484
|
Cytiva
SH30802.02 |
Without L-Glutamine | 1000 mL bottle |
£102.00
1000mL |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Increase process yields for the manufacture of monoclonal antibodies for therapeutic use with HyClone™ CDM4MAb™ Media, a chemically-defined media containing no animal derived components.
Specifications
2° to 8°C, away from light | |
Liquid | |
CDM4MAb Liquid Medium | |
0.2 μm Sterile-filtered |