missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ CDM4PERMAb Powdered Cell Culture Media Without L-glutamine
Chemically defined ADCF media
Brand: Cytiva SH30872.02
Additional Details : Weight : 0.01000kg
Description
- For use with PER.C6 cells
- Animal-derived component-free
- Chemically defined formulation
- Designed for high cell yield and recombinant protein production
- Allows for direct or sequential adaption
- Component traceability
- Manufactured according to cGMP guidelines
- Tested for fed-batch bioreactor cultures
Specifications
2 to 8 °C away from light and moisture | |
Powder | |
Non-sterile | |
No Glutamine | |
10 L |
PER.C6 | |
HyClone Cell Culture Media | |
Phenol Red | |
For high cell yield and recombinant protein production |