Learn More
Cytiva HyClone™ CDM4PERMAb Powdered Media
Chemically defined ADCF media for use with PER.C6 cells.
Brand: Cytiva SH30872.01
Code : 83
Additional Details : CAS Number : 7758-99-8 Weight : 0.10300kg
Description
HyClone CDM4PERMAb medium has a chemically defined and animal-derived component-free (ADCF) formulation. This high-performance cell culture medium has been developed to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 technology.
The medium has been successfully tested in a variety of applications, including fed-batch bioreactor cultures.
CDM4PERMAb is available in liquid and powder formats in user-friendly packaging.
- Animal-derived component-free
- Chemically defined formulation
- Designed for high cell yield and recombinant protein production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured according to cGMP guidelines
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Phenol Red |