missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cell Boost™ 3 Supplement
Fed-batch supplement with amino acids, vitamins, glucose, and trace elements for hybridoma and myeloma cells
£107.00 - £118.00
Specifications
Cell Type | Hybridomas, Myelomas |
---|---|
Form | Liquid |
Product Type | Cell Boost Supplement |
Format | Bottle |
For Use With (Application) | Improving Productivity and Cell Growth in Fed-Batch Cultures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16524581
|
Cytiva
SH31115.01 |
500 mL |
£107.00
500mL |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16675795
|
Cytiva
SH31115.02 |
1000 mL |
£118.00
1 L |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
- Supports fed-batch cultures with amino acids, vitamins, glucose, and trace elements
- Developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
Hybridomas, Myelomas | |
Cell Boost Supplement | |
Improving Productivity and Cell Growth in Fed-Batch Cultures |
Liquid | |
Bottle | |
HyClone™ |