missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cell Boost™ 4 Supplement
Fed-batch media supplement containing amino acids, vitamins, glucose, trace elements, and growth factors for CHO cells
£121.00 - £172.00
Specifications
Cell Line | CHO |
---|---|
Form | Liquid |
Product Type | Cell Boost Supplement |
Format | Bottle |
For Use With (Application) | Improving Productivity and Cell Yield in Fed-Batch Cultures |
Product Code | Brand | Reconstituted Volume | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Reconstituted Volume | Price | Quantity & Availability | |||||
16534581
|
Cytiva
SH31116.01 |
500 mL |
£121.00
500mL |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16685795
|
Cytiva
SH31116.02 |
1000 mL |
£172.00
1 L |
Estimated Shipment: 27-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
- Supports fed-batch cultures with amino acids, vitamins, glucose, trace elements, and growth factors
- Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
CHO | |
Cell Boost Supplement | |
Improving Productivity and Cell Yield in Fed-Batch Cultures |
Liquid | |
Bottle | |
HyClone™ |