Learn More
Cytiva HyClone™ Cell Boost™ 5 Supplement
Fed-batch supplement with lipids, amino acids, vitamins, and growth factors for hybridoma, HEK 293, NS0, and CHO cells
Brand:  Cytiva SH31117.01
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 16544581
  • £110.00 / 500mL
Estimated Shipment: 09-08-2024
to see stock.
Description
Description
  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
  • Supports fed-batch cultures with lipids, amino acids, vitamins, and growth factors
  • Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line, Human Embryonic Kidney (HEK) 293, NS0, and hybridoma cells
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
Specifications
Specifications
CHO, HEK293, Hybridoma, NS0
Cell Boost Supplement
Improving Productivity and Cell Yield in Fed-Batch Cultures
500 mL
Liquid
Bottle
HyClone™