Learn More
Cytiva HyClone™ Cell Boost™ 6 Supplement
Fed-batch supplement with lipids, amino acids, vitamins, and growth factors for hybridoma, HEK 293, NS0, CHO, and T cells
Brand: Cytiva SH31118.02
Additional Details : Weight : 1.06000kg
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
- Supports fed-batch cultures with lipids, amino acids, vitamins and growth factors
- Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line, Human Embryonic Kidney (HEK) 293, NS0, hybridoma, and T cells
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
CHO, HEK293, Hybridoma, NS0, T cells | |
Cell Boost Supplement | |
Improving Productivity and Cell Yield in Fed-Batch Cultures | |
1000 mL |
Liquid | |
Bottle | |
HyClone™ |