Learn More
Cytiva HyClone™ Cell Boost™ 7a Supplement
A ready-to-use, powder media supplement optimized for high-yield recombinant protein production in CHO fed-batch processes
Brand: Cytiva SH31119.01
Additional Details : Weight : 0.50000kg
Description
- Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
- Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
- Designed to support modern manufacturing processes
- Chemically defined (CD), animal-derived component-free (ADCF), and free of any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
- Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
- Manufactured to meet cGMP manufacturing standards and QC specifications
Specifications
Without: L-glutamine, With: Poloxamer 188 | |
Liquid | |
Non-sterile | |
HyClone™ |
CHO | |
Cell Culture Supplement | |
High-Yield Recombinant Protein Production in CHO Fed-Batch Processes | |
0.5 L |