Learn More
Cytiva HyClone™ Cell Boost 7a Supplement
Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose. Manufactured to meet cGMP manufacturing standards and QC specifications.
£621.00 - £936.00
Specifications
Content And Storage | 2 to 8°C, away from light and moisture |
---|---|
Form | Powder |
Sterility | Non-sterile |
With Additives | Poloxamer 188 |
Without Additives | No Glutamine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15805261
|
Cytiva
SH31026.07 |
1 L |
£621.00
Each |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952447
|
Cytiva
SH31026.01 |
5 L |
£936.00
5 L |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines.
Cell Boost 7a is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.
- Designed to support modern manufacturing processes, Cell Boost 7a is chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
- For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
No Glutamine |
Powder | |
Poloxamer 188 |