Learn More
Cytiva HyClone™ Cell Boost™ 7b Supplement
A concentrated amino acid supplement, in a ready to use powder format, for high-yield protein production in CHO fed-batch processes
Brand: Cytiva SH31120.01
73.70 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
- A concentrated solution of amino acids with an alkaline pH
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production.
- Designed to support modern manufacturing processes
- Chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
- Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
- Manufactured to meet cGMP manufacturing standards and QC specifications
Specifications
Without: L-glutamine, poloxamer 188, and insulin | |
Amino Acid Supplement | |
HyClone™ |
Liquid | |
High-Yield Protein Production in CHO Fed-Batch Processes | |
100 mL |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.