Learn More
Cytiva HyClone™ Cell Boost 7b Supplement
Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.
£112.00 - £473.00
Specifications
Content And Storage | 2 to 8°C, away from light and moisture |
---|---|
Form | Powder |
Sterility | Non-sterile |
Without Additives | No Glutamine, No Poloxamer 188, No Insulin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815261
|
Cytiva
SH31027.08 |
0.1 L |
£112.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16457695
|
Cytiva
SH31027.07 |
1 L |
£473.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines.
Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.
- Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pH. Supplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
- For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
Specifications
2 to 8°C, away from light and moisture | |
Powder | |
Non-sterile | |
No Glutamine, No Poloxamer 188, No Insulin |
Safety and Handling
missing translation for 'shelfLife' : 18 months
- Cell Boost 7b, without Poloxamer-188, without Insulin, without L-Glutamine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.