missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin

Sterile-filtered and quality-tested to ensure dependable, reproducible cell growth

Brand:  Cytiva SH30084.03IR30-40

 View more versions of this product

Product Code. 16211102

  • £1160.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Meets the requirements of most users
  • Filtered through triple 100 nm pore size-rated filters
  • Standard endotoxin and hemoglobin specifications
  • Subjected to moderate biochemical analyses
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certificate of Suitability No. R1-CEP 2000-384
Serum
500 mL
Characterized
≤ 25 EU/mL
HyClone™
Australia
Fetal Bovine Serum
Sequential Triple-filtered, 100 nm
Irradiated (30-40 kGy)
≤ 25 mg/dL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers