missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin

Sterile-filtered and quality-tested to ensure dependable, reproducible cell growth

£121.38 - £1160.00

Specifications

Certifications/Compliance Certificate of Suitability No. R1-CEP 2000-384
Country of Origin Australia
Form Serum
Product Type Fetal Bovine Serum
Sterilization Method Sequential Triple-filtered, 100 nm
View More Specs

Products 7
Product Code Brand Quantity Processing Price Quantity & Availability  
Product Code Brand Quantity Processing Price Quantity & Availability  
16645745
View Documents
Cytiva
SH30084.01
50 mL None This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16655745
View Documents
Cytiva
SH30084.02IH
100 mL Heat Inactivated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16211102
View Documents
Cytiva
SH30084.03IR30-40
500 mL Irradiated (30-40 kGy)
£1160.00
500mL
Please to purchase this item. Need a web account? Register with us today!
16665745
View Documents
Cytiva
SH30084.03E
500 mL Embryonic Stem Cell Screened This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16675745
View Documents
Cytiva
SH30084.03M
500 mL Mesenchymal Stem Cell Screened This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16685745
View Documents
Cytiva
SH30084.50E
50 mL Embryonic Stem Cell Screened This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16695745
View Documents
Cytiva
SH30084.50M
50 mL Mesenchymal Stem Cell Screened This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Meets the requirements of most users
  • Filtered through triple 100 nm pore size-rated filters
  • Standard endotoxin and hemoglobin specifications
  • Subjected to moderate biochemical analyses
Specifications

Specifications

Certificate of Suitability No. R1-CEP 2000-384
Serum
Sequential Triple-filtered, 100 nm
≤ 25 EU/mL
HyClone™
Australia
Fetal Bovine Serum
Characterized
≤ 25 mg/dL
Videos
SDS
Documents

Documents

Product Certifications