missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin
Enhance growth of cells with New Zealand sourced HyClone Characterized Fetal Bovine Serum.
Brand: Cytiva SH30406.02
759.27 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
- The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
- One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
- Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
- Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
Specifications
Progesterone Chloride Insulin Inorganic Phosphorus PTH Glucose Cholesterol Copper HDL Magnesium LDL Selenium Triglycerides IgG Gamma Globulin pH Phospholipids Osmolality Alkaline Phosphatase Iron Lactate Dehydrogenase TIBC SGPT Percent Saturation SGOT B1 B2 B6 Niacin Biotin Retinol Carotene Tocopherol B12 Folate Vitamin C 1,25-dihydroxy vitamin D Total Protein Albumin Blood Urea Nitrogen | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C | |
Sterile | |
None | |
≤ 20 EU/mL | |
Yes |
USDA Regulations | |
500 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 25 mg/dL | |
USDA Inspected |