missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin
Low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells.
£141.00 - £974.10
Specifications
Country of Origin | United States |
---|---|
Form | Serum |
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
16615735
|
Cytiva
SH30071.02IH |
Heat-Inactivated, Irradiated | 100 mL | N/A | |||||
16261082
|
Cytiva
SH30071.03IH25-40 |
Heat Inactivated, Irradiated (25-40 kGy) | 500 mL |
£699.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16271082
|
Cytiva
SH30071.03IH30-45 |
Heat Inactivated, Irradiated (30-45 kGy) | 500 mL |
£638.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16625735
|
Cytiva
SH30071.02IR |
Irradiated | 100 mL | N/A | |||||
16635735
|
Cytiva
SH30071.03IR35-50 |
Irradiated | 500 mL | N/A | |||||
16645735
|
Cytiva
SH30071.04 |
None | 1000 mL | N/A | |||||
Description
- Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
- Low in antibodies and high in growth factors
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Each lot is subjected to virus panel testing according to 9 CFR 113.53
- Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
- Supports robust growth and division of cells, including embryonic stem cells
- Produced using true pool technology to minimize bottle-to-bottle variability
- Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
- Sourced in the USA
Specifications
United States | |
Fetal Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
Virus Panel Testing According to 9 CFR 113.53 |
Serum | |
Sterile | |
Characterized | |
HyClone™ |