Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)
Enhance growth of cells with New Zealand sourced Characterized FBS.
Brand:  Cytiva SH30406.02IR25-40
 View more versions of this product
Product Code. 16271112
  • £985.00 / 500mL
Explore more special offers
Description
Description
  • Carefully collected, processed, and filtered in our own New Zealand facility
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • Greater security against bovine disease, since New Zealand has the fewest reported bovine diseases in the world
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles using true pool technology
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen
    500 mL
    Sequential-filtered, 100 nm
    Irradiated, 25 to 40 doses
    ≤ 20 EU/mL
    ≤ 25 mg/dL
    USDA Inspected
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Sterile
    Characterized
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    No
    Yes