missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

Brand:  Cytiva SH30071.03IR

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11306060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
Specifications

Specifications

Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Percent Saturation, Creatinine, Potassium, Glucose, SGPT, Sodium, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
HyClone™
USDA Inspected
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
United States
500 mL
Sequential Triple-filtered, 100 nm
Irradiated
≤ 25 EU/mL
≤ 25 mg/dL
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers