missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

Brand:  Cytiva SH30071.03HI

657.77 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 12379802

  • £1060.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
500 mL
Sequential Triple-filtered, 100 nm
Heat Inactivated
≤ 25 EU/mL
≤ 25 mg/dL
No
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Yes
HyClone™
USDA Inspected
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers