missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

Brand:  Cytiva SH30071.02IH25-40

 View more versions of this product

Product Code. 17296886

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Albumin
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Liquid
100 mL
Sequential Triple-filtered, 100 nm
Heat Inactivated, Irradiated (25 to 40 doses)
≤ 25 EU/mL
≤ 25 mg/dL
No
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
United States
Fetal Bovine Serum
Sterile
Characterized
Bluetongue
Yes
HyClone™
USDA Inspected
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers