missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum, EMEA Tested
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
Brand: Cytiva SH30533.04NI
Additional Details : Weight : 1.15000kg
Description
- High-Quality Bovine Calf Serum
- Fortified with iron and naturally derived components
- Augmented with additional growth-promoting components to yield an exceptional bovine calf serum
- Excellent performance with a variety of cell types
Specifications
Calf Bovine Serum | |
Sterile | |
Non-Irradiated | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |
1000 mL | |
Sequential Triple-filtered, 100 nm | |
HyClone™ |