Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
Brand:  Cytiva SH30087.04IR25-40
 View more versions of this product
Product Code. 16241102
  • £217.00 / 1000mL
Explore more special offers
Description
Description
  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
1000 mL
Sequential-filtered, 100 nm, Gamma-irradiated
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Iron and naturally derived components
Sterile
Irradiated, 25 to 40 doses
No