missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
Brand: Cytiva SH30087.04IR30-45
192.85 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Age at time of collection: typically 16 - 22 weeks
- Iron-supplemented with additional growth promoting components
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
1000 mL | |
Sequential-filtered, 100 nm, Gamma-irradiated | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Iron and naturally derived components | |
Sterile | |
Irradiated, 30 to 45 doses | |
No |