missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
£39.35 - £241.00
Specifications
Additional Assays | Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG |
---|---|
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12349842
|
Cytiva
SH30087.02HI |
100 mL |
£39.35
100mL |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12694227
|
Cytiva
SH30087.04HI |
1000 mL |
£241.00
1000mL |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.
Specifications
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
Sequential-filtered, 100 nm, Gamma-irradiated | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Sterile | |
Heat Inactivated | |
Yes |