missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Adult Bovine Serum (U.S.)
Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).
Brand: Cytiva SH30075.05IR30-45
This item is not returnable.
View return policy
Description
- Sourced from a closed herd maintained on USDA-licensed premises
- Cattle from these closed herds are used exclusively for the production of donor bovine blood products
Specifications
Certificate of Suitability No. R1-CEP 2001-259 | |
Liquid | |
1 L | |
Irradiated (30 to 45 doses) | |
No | |
HyClone™ |
United States | |
Adult Bovine Serum | |
Sequential Triple-filtered, 100 nm | |
≤ 50 EU/mL | |
≤ 25 mg/dL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction