missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Donor Equine Serum
Carefully collected from selected U.S. donor herds that receive regular veterinary inspection and care.
£103.00 - £111.00
Specifications
Additional Assays | Albumin |
---|---|
Country of Origin | United States |
Product Type | Equine Serum |
Quantity | 1 L |
Sterility | Sterile |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
11516541
|
Cytiva
SH30074.04 |
1 L | None |
£111.00
Each |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17286936
|
Cytiva
SH30074.04IR |
1 L | Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Serum Collection:- From collection to final packaging
- Every step is documented
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Aseptically packaged in a Class 100 clean room condition
- Each lot of serum is tested to ensure quality and sterility
- Pooled after filtration and before final packaging
- Consistent quality from the first bottle filled to the last
- Increases consistency and minimizes variability with a lot of serum
Specifications
Albumin | |
Equine Serum | |
Sterile | |
No |
United States | |
1 L | |
Sequential Triple-filtered, 100 nm | |
HyClone™ |