missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum (U.S.)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.
£14.75 - £74.45
Specifications
Additional Assays | Albumin, Lactate Dehydrogenase, Osmolality, Percent Saturation, Chloride, Creatinine, SGOT, SGPT, Sodium, Iron, Total Bilirubin, Alkaline Phosphatase, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron |
---|---|
Country of Origin | United States |
Form | Liquid |
Product Type | Equine Serum |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Processing | Price | Quantity & Availability | |||
11581831
|
Cytiva
SH30074.02 |
Donor Equine Serum (U.S.) | 100 mL | None |
£14.75
100mL |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12319822
|
Cytiva
SH30074.03HI |
Donor Equine Serum (U.S.), Heat Inactivated | 500 mL | Heat Inactivated |
£74.45
500mL |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11591831
|
Cytiva
SH30074.03 |
Donor Equine Serum (U.S.) | 500 mL | None |
£56.55
500mL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Format: Liquid
Specifications
Albumin, Lactate Dehydrogenase, Osmolality, Percent Saturation, Chloride, Creatinine, SGOT, SGPT, Sodium, Iron, Total Bilirubin, Alkaline Phosphatase, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron | |
Liquid | |
Sequential Triple-filtered, 100 nm |
United States | |
Equine Serum | |
BVDV, Cytopathogenic Agents, Rabies Virus, Reovirus |