missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
£27.90 - £74.45
Specifications
Additional Assays | Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG |
---|---|
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
17246896
|
Cytiva
SH30074.03IR25-40 |
Irradiated (25 to 40 kGy) | 500 mL |
£74.45
1 L |
Estimated Shipment: 15-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12694207
|
Cytiva
SH30074.02HI |
Heat Inactivated | 100 mL |
£27.90
100mL |
Estimated Shipment: 15-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12339822
|
Cytiva
SH30074.03IR |
Irradiated | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Specifications
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
Sequential-filtered, 100 nm |