missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum, US Origin
Click to view available options
Processing:
Heat Inactivated
Irradiated (25 to 40 kGy)
Unit Size:
1 L
Description
- Carefully collected from animals fed an enriched diet to ensure proper nutrition
- To reduce lipid concentration, the equine herd is fasted and then the blood is collected in a special facility using aseptic techniques
- Coggins tests for equine infectious anemia are performed on each horse
Specifications
Specifications
| Age | >6 months |
| Country of Origin | United States |
| Form | Liquid |
| Product Type | Equine Serum |
| Quantity | 1000 mL |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Processing | Heat Inactivated |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | Yes |
| Hemoglobin Concentration | ≤ 10 mg/dL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction