missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ DPM-PF-CHO Powdered Media
Powder mediums (Main/Base) used to create PF-CHO MPS media
Brand: Cytiva SH30335.01
Additional Details : Weight : 0.06620kg
Description
- Designed for high cell yield and recombinant protein production
- Allow for direct or sequential adaptation
- Support growth in multiple cell culture systems
- Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
Specifications
2 to 8°C, away from light and moisture | |
Powder | |
No Glutamine, No Phenol Red, No Poloxamer 188 |
Base powdered media | |
Non-sterile | |
5 L |