missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Modified Eagles Medium

Designed to support cell culture growth and biomanufacturing processes.

Brand:  Cytiva SH30585.02

21.76 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10658364

  • £25.30 / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 9001:2000
Liquid modified medium with glucose; without phenol red, sodium pyruvate, and L-glutamine; 1000 mL bottle
DMEM/High glucose modified without phenol red, sodium pyruvate, L-glutamine
Glucose
1000 mL
2° to 8°C, away from light
Liquid
0.1 μm Sterile-filtered
No Glutamine, No Phenol Red, No Sodium Pyruvate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers