missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder
£15.90 - £58.80
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Content And Storage | 2° to 8° C |
Form | Powder |
Sterility | Non-sterile |
Without Additives | No Calcium, No Magnesium |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11511801
|
Cytiva
SH30013.02 |
Without Calcium, Magnesium (Powder), 2 x 5L |
£19.90
Pack of 2 |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10021342
|
Cytiva
SH30013.03 |
Without Calcium, Magnesium (Powder), 10L |
£15.90
10 L |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10708334
|
Cytiva
SH30013.04 |
Without Calcium, Magnesium (Powder), 50L |
£58.80
50 L |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Powder | |
No Calcium, No Magnesium |
2° to 8° C | |
Non-sterile |