Learn More
Cytiva HyClone™ Earle's 1X Balanced Salt Solution
Liquid buffer solution membrane-sterilized through 0.1μm pore filters for use in cell culture applications. Cytiva HyClone™ Earle's 1X Balanced Salt Solution incorporates a rigorous specification process, uses traceable ingredients, and is manufactured according to cGMP guidelines.
Brand: Cytiva SH30029.02
15.30 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Code : A0
Additional Details : Weight : 0.50000kg
Description
Components:- Calcium chloride (anhydrous): 200 mg/L
- Potassium chloride: 400 mg/L
- Potassium phosphate monobasic (anhydrous): 0 mg/L
- Magnesium sulfate (anhydrous): 97.67 mg/L
- Sodium chloride: 6800 mg/L
- Sodium phosphate dibasic (anhydrous): 0 mg/L
- Sodium phosphate monobasic H20: 140 mg/L
- D-Glucose (anhydrous): 1000 mg/L
- Phenol red-Na: 11 mg/L
- Sodium bicarbonate: 2200 mg/L
Specifications
cGMP and ISO 9001:2000 Guidelines | |
1 X, With Calcium, Magnesium and Phenol Red | |
Liquid | |
0.1 μm Sterile-filtered | |
=1.0EU/mL | |
500 mL |
15° to 3°C | |
1 X | |
Sterile | |
Calcium, Magnesium, Phenol Red | |
Cell Culture |