Learn More
Cytiva HyClone™ Fetal Bovine Serum, South American Origin
Brand: Cytiva SV30160.03
290.85 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.54000kg
Description
- Filtered through three sequential 100nm (0.1μm) pore-size rated filters
- Not sold in the U. S. Only for sale to European and Asian customers