missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
£88.55 - £650.00
Specifications
Country of Origin | United States |
---|---|
Specialty Type | Charcoal/Dextran filtered |
Sterility | Sterile |
Grade | Specialty filtered |
Endotoxin Concentration | ≤ 10 EU/mL |
Product Code | Brand | Quantity | Processing | Heat Inactivated | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Heat Inactivated | Price | Quantity & Availability | |||
12339782
|
Cytiva
SH30068.03IR |
500 mL | Gamma Irradiated | No | |||||
10611235
|
Cytiva
SH30068.03 |
500 mL | None | No |
£635.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12329782
|
Cytiva
SH30068.03HI |
500 mL | Heat Inactivated | Yes |
£650.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12319782
|
Cytiva
SH30068.02HI |
100 mL | Heat Inactivated | Yes |
£166.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12634217
|
Cytiva
SH30068.01HI |
50 mL | Heat Inactivated | Yes |
£105.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12389772
|
Cytiva
SH30068.01 |
50 mL | None | No |
£88.55
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11571821
|
Cytiva
SH30068.02 |
100 mL | None | No |
£151.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16201082
|
Cytiva
SH30068.03IR25-40 |
500 mL | Gamma Irradiated, 25 to 40 doses | No |
£650.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
- This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
- Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
- Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
United States | |
Sterile | |
≤ 10 EU/mL | |
No |
Charcoal/Dextran filtered | |
Specialty filtered | |
≤ 20 mg/dL | |
USDA Inspected |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-2.1 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 2.1% of components with unknown hazards to the aquatic environment.