missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Brand: Cytiva SH30070.03IR30-45
713.26 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40 nm pore-size rated filters
Specifications
25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1 | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Fetal Bovine Serum | |
Sterile | |
Defined | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
HyClone™ | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
500 mL | |
Sequential-filtered, 40 nm | |
Irradiated (30 to 45 doses) | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
No |