missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard

Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)

Brand:  Cytiva SH30088.03

411.84 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11521851

  • £487.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Support cell culture growth
  • Standard grade
  • Aseptically packaged in Class 100 cleanroom
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
United States
500 mL
Sequential-filtered, 100 nm
None
For Information Only
For Information Only
USDA Inspected
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Standard Fetal Bovine Serum, US Origin
Fetal Bovine Serum
Sterile
Standard
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers