missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, USDA Tested
Sourced from countries recognized as being free of BSE and FMD by the USDA
£97.00 - £463.00
Specifications
Certifications/Compliance | USDA Regulations |
---|---|
Description | USDA Tested Fetal Bovine Serum, Central American Sourced |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Grade | Characterized |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
17246916
|
Cytiva
SH30910.02 |
100 mL | None | N/A | |||||
17266916
|
Cytiva
SH30910.03 |
500 mL | None | N/A | |||||
17256916
|
Cytiva
SH30910.02HI |
100 mL | Heat Inactivated | N/A | |||||
17276916
|
Cytiva
SH30910.03HI |
500 mL | Heat Inactivated | N/A | |||||
Description
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Specifications
USDA Regulations | |
Sterile | |
Characterized | |
USDA Inspected |
USDA Tested Fetal Bovine Serum, Central American Sourced | |
Sequential-filtered, 100 nm | |
No |