missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ Fetal Bovine Serum, USDA Tested
Sourced from countries recognized as being free of BSE and FMD by the USDA
Brand: HyClone products Cytiva SH30910.03IR
This item is not returnable.
View return policy
Description
- USDA safety-tested
- Filtered through three sequential 100nm (0.1μm) pore-size rated filters
Additional assays performed (subject to change without notice): Alkaline Phophatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Glucose, Gamma Globulin, Electrophoresis, and IgG.
Specifications
Albumin | |
<-1°C | |
Fetal Bovine Serum | |
Sterile | |
Central America | |
≤25EU/mL | |
≤25mg/dL | |
USDA Inspected |
USDA | |
Central America | |
500 mL | |
100 nm filtration, Irradiated | |
Characterized | |
No | |
No |