missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Clone I Serum
HyClone™ FetalClone™ I is a fetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
£310.00 - £368.00
Specifications
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG |
---|---|
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Country of Origin | United States |
Quantity | 500 mL |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
16291092
|
Cytiva
SH30080.03IR25-40 |
500 mL | Irradiated (25 to 40 doses) |
£368.00
500mL |
Estimated Shipment: 10-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11940958
|
Cytiva
SH30080.03HI |
500 mL | Heat Inactivated |
£362.00
500mL |
Estimated Shipment: 10-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12359832
|
Cytiva
SH30080.03IR |
500 mL | Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
500 mL | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
≤ 20 mg/dL | |
No |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
Sterile | |
≤ 10 EU/mL | |
Typically less than 2 μg/mL | |
USDA Inspected |