missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ I Serum (U.S.)
Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.
Brand: Cytiva SH30080.02HI
95.95 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
FBS Replacement Serum | |
Sterile | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
No |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
100 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
Typically less than 2μg/mL | |
USDA Inspected |