missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Product Code. p-7145851
Click to view available options
Quantity:
100 mL
500 mL
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 12339772

Brand: Cytiva SH30066.02HI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for the growth of CHO cells and derivatives.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 100 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing Heat Inactivated
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated Yes
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2 μg/mL
    Product Line HyClone
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.