missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ II Serum, US Origin
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Beskrivning
- A supplement used to enrich cell culture performance
- Delivers both cost-effectiveness and high performance
- Commonly used in cell culture and bioprocessing applications
- Low endotoxin and hemoglobin levels
- High lot-to-lot consistency
Specifikationer
Specifikationer
| Certifications/Compliance | Manufactured using ISO 9001- and ISO 13485-Certified Processes |
| Content And Storage | Store at or below -10°C |
| Country of Origin | United States |
| Form | Liquid |
| Product Type | FBS Replacement Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Processing | Heat Inactivated, Irradiated |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine Respiratory Syncytial Virus, Bovine Viral Diarrhea Virus, Rabies, and Reovirus |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering