missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ III Serum (U.S.)

Product Code. p-7058046
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
None
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10295162

Brand: Cytiva SH30109.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Country of Origin United States
Product Type FBS Replacement Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 20 mg/dL
IgG levels Typically less than 2 μg/mL
Product Line HyClone
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.