Learn More
Cytiva HyClone™ HEPES Solution
Maintains physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration). HyClone™ HEPES Solution is part of high-performance line of nutritional and growth supplements.
Brand: Cytiva SH30237.01
16.77 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 0.13500kg
Description
- Format: Liquid
Specifications
Manufactured under cGMP and ISO 9001:2000 guidelines | |
Liquid | |
0.1 μm Sterile-filtered | |
100 mL |
2° to 8°C | |
HEPES Buffer | |
7.2 to 7.5 |