Learn More
Cytiva HyClone™ Human Mesenchymal Stem Cell Screened Characterized Fetal Bovine Serum, US Origin
Designed for human mesenchymal stem cells
Brand: Cytiva SH30071.03M
This item is not returnable.
View return policy
Description
Human Mesenchymal Stem Cell Screened FBS supports the growth of undifferentiated human mesenchymal stem cells (hMSC). During screening, hMSC are observed for evidence of nutritional deficiency, cytotoxicity, or morphological aberrations.Specifications
United States | |
Human mesenchymal stem cell screened | |
40 nm filtration | |
No |
Liquid | |
500 mL | |
≤ 10 EU/mL | |
≤ 10 mg/dL |