missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ HyCell CHO Powder Cell Culture Medium without L-glutamine and with HT

Product Code. 12872986
Click to view available options
Quantity:
10 L
1000 mL PETE bottle
5 L
Unit Size:
10 L
1000mL
5 L
This item is not returnable. View return policy

Product Code. 12872986

Brand: Cytiva SH30933.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A versatile, animal derived component free (ADCF) cell culture media.

  • Chemically defined and animal-derived component‑free formulations
  • Designed for high cell yield and recombinant protein production
  • Allow direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • Developed for Chinese hamster ovary (CHO) cell culture in bioprocessing applications
  •  Specifically formulated to provide excellent performance, support high peak cell density, and enhance product yield and quality
  • Can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms ranging from shake flasks to large-scale bioreactors
  • TRUSTED_SUSTAINABILITY

    Specifications

    Form Powder
    Sterility Non-sterile
    With Additives Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine
    Without Additives No Phenol Red, No HEPES, No Glutamine
    Quantity 5 L
    Content And Storage 2° to 8°C, away from light and moisture
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.