missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ HyCell CHO Powder Cell Culture Medium without L-glutamine and with HT

A versatile, animal derived component free (ADCF) cell culture media.

Brand:  Cytiva SH30933.02

393.63 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 0.28260kg

 View more versions of this product

Product Code. 17602085

  • £466.00 / 10 L
Estimated Shipment: 06-09-2024
to see stock.

Description

Description

  • Chemically defined and animal-derived component‑free formulations
  • Designed for high cell yield and recombinant protein production
  • Allow direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • Developed for Chinese hamster ovary (CHO) cell culture in bioprocessing applications
  •  Specifically formulated to provide excellent performance, support high peak cell density, and enhance product yield and quality
  • Can be used for batch, fed-batch, or perfusion applications in multiple cell culture platforms ranging from shake flasks to large-scale bioreactors
  • Specifications

    Specifications

    2° to 8°C, away from light and moisture
    Non-sterile
    No Phenol Red, No HEPES, No Glutamine
    Powder
    Sodium Bicarbonate, Poloxamer 188, Hypoxanthine, Thymidine
    10 L
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers